(S)-(+)-2-Pentanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(+)-Propyl methyl carbinol, (S)-(+)-Methyl propyl carbinol, (S)-(+)-2-Hydroxy pentane

P0743-1ML P0743-5ML
TCP0743-1MLEA 161 USD
TCP0743-1ML TCP0743-5ML
(S)-(+)-2-Pentanol ≥98.0% (by GC)
(S)-(+)-2-Pentanol

CAS Number: 26184-62-3
MDL Number: MFCD00065952
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 34
Specific Gravity (20/20): 0.81

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 119 °C
Melting Pt: -50
Density: 0.81 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00065952
CAS Number: 26184-62-3
UN: 1105
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c90b6faf4fa2e709c6b9b78da772d06df7d30da&vid=9c90b6faf4fa2e709c6b9b78da772d06df7d30da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR