Phenolphthalein 0,04% in ethanol 40%
Supplier: TCI America
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
P0702-100ML
P0702-500ML
TCP0702-100MLEA
34.32
USD
TCP0702-100ML
TCP0702-500ML
Phenolphthalein 0,04% in ethanol 40%
Phenolphthalein
[for pH Determination]
CAS Number: 77-09-8
MDL Number: MFCD00005913
Molecular Weight: 318.33
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 24
Formula:
C₂₀H₁₄O₄ Flash Pt: 24 °C |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...