3-Phenylpropylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Phenyl-1-propylamine

P0664-25G P0664-5G
TCP0664-25GEA 78.36 USD
TCP0664-25G TCP0664-5G
3-Phenylpropylamine ≥97.0% (by GC, titration analysis)
3-Phenylpropylamine

CAS Number: 2038-57-5
MDL Number: MFCD00008224
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 90
Specific Gravity (20/20): 0.95

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 220 °C
Density: 0.95 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00008224
CAS Number: 2038-57-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7540b3e8f5caf548c3531a3a1b057f3f9d6eebae&vid=7540b3e8f5caf548c3531a3a1b057f3f9d6eebae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR