Catechol violet

Supplier: TCI America

Synonyms: Pyrocatechol Violet

P0568-1G P0568-5G
TCP0568-1GEA 25 USD
TCP0568-1G TCP0568-5G
Catechol violet
Catechol violet

CAS Number: 115-41-3
MDL Number: MFCD00005868
Molecular Formula: C19H14O7S
Molecular Weight: 386.37
Form: Crystal
Color: Deep Yellow Red

Formula: C₁₉H₁₄O₇S
MW: 386.38 g/mol
MDL Number: MFCD00005868
CAS Number: 115-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ccecb00b27d614e8c973b429a60fc5a574a3d02&vid=0ccecb00b27d614e8c973b429a60fc5a574a3d02&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR