Propionaldehyde ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Propanal
P0498-25ML
P0498-500ML
TCP0498-25MLEA
27.29
USD
TCP0498-25ML
TCP0498-500ML
Propionaldehyde ≥98.0% (by GC)
Propionaldehyde
CAS Number: 123-38-6
MDL Number: MFCD00007020
Molecular Formula: C3H6O
Molecular Weight: 58.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 46
Melting point (°C): -81
Flash Point (°C): -37
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: Propanal
Formula:
C₃H₆O MW: 58.08 g/mol Boiling Pt: 46 °C Melting Pt: -81 °C Density: 0.81 (20 °C) Flash Pt: -37 °C |
MDL Number:
MFCD00007020 CAS Number: 123-38-6 UN: 1275 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...