Pivalic acid ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 2,2-Dimethylpropionic acid, Trimethylacetic acid

P0461-25G P0461-400G
TCP0461-25GEA 23 USD
TCP0461-25G TCP0461-400G
Pivalic acid ≥99.0% (by GC)
Pivalic acid

CAS Number: 75-98-9
MDL Number: MFCD00004194
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 164
Flash Point (°C): 64
Freezing point (°C): 34


Other Product Names / Synonyms: 2,2-Dimethylpropionic Acid, Trimethylacetic Acid

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 164 °C
Flash Pt: 64 °C
MDL Number: MFCD00004194
CAS Number: 75-98-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b4ebfd064035db1129782e4e709826f039a82a&vid=11b4ebfd064035db1129782e4e709826f039a82a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR