trans-2-Pentenoic acid ≥95.0% (by GC)

Supplier: TCI America

P0345-25ML P0345-5ML
TCP0345-25MLEA 150 USD
TCP0345-25ML TCP0345-5ML
trans-2-Pentenoic acid ≥95.0% (by GC)
trans-2-Pentenoic acid

CAS Number: 13991-37-2
MDL Number: MFCD00002704
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 102
Specific Gravity (20/20): 0.99

Formula: C₅H₈O₂
Boiling Pt: 97 °C
Density: 0.99 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00002704
CAS Number: 13991-37-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e046d31f98eecfe6297daad3020c5e8e777d293e&vid=e046d31f98eecfe6297daad3020c5e8e777d293e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR