4-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Phenyl-3-thiosemicarbazide, N-Phenylhydrazinecarbothioamide, 4-Phenylthiosemicarbazide

P0235-1G P0235-25G
TCP0235-1GEA 21 USD
TCP0235-1G TCP0235-25G
4-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)
4-Phenyl-3-thiosemicarbazide

CAS Number: 5351-69-9
MDL Number: MFCD00007615
Molecular Formula: C7H9N3S
Molecular Weight: 167.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 140

Formula: C₇H₉N₃S
MW: 167.23 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00007615
CAS Number: 5351-69-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR