1,10-Phenanthroline monohydrate ≥99.0%

Supplier: TCI America

Synonyms: 1,10-Phenanthroline hydrate

P0221-1G P0221-25G
TCP0221-1GEA 28 USD
TCP0221-1G TCP0221-25G
1,10-Phenanthroline monohydrate ≥99.0%
1,10-Phenanthroline monohydrate

CAS Number: 5144-89-8
MDL Number: MFCD00149973
Molecular Formula: C12H8N2
Molecular Weight: 180.21
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 102

Formula: C₁₂H₈N₂·1H₂O
Melting Pt: 102 °C
MDL Number: MFCD00149973
CAS Number: 5144-89-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd137c09faaabf217e3954eecb871a145f96ecf2&vid=cd137c09faaabf217e3954eecb871a145f96ecf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR