p-Phenylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,4-Diaminobenzene, PPD, 1,4-Phenylenediamine

P0170-250G P0170-25G
TCP0170-250GEA 73.97 USD
TCP0170-250G TCP0170-25G
p-Phenylenediamine ≥98.0% (by GC, titration analysis)
p-Phenylenediamine

CAS Number: 106-50-3
MDL Number: MFCD00007901
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 142
Flash Point (°C): 152


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,4-Diaminobenzene

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 267 °C
Melting Pt: 142 °C
Flash Pt: 152 °C
MDL Number: MFCD00007901
CAS Number: 106-50-3
UN: 1673
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b11c648a074c66f0416f17e0cc280af78817be64&vid=b11c648a074c66f0416f17e0cc280af78817be64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR