1,8-Octanedithiol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,8-Dimercaptooctane
O0374-5G
O0374-1G
TCO0374-5GEA
103
USD
TCO0374-5G
TCO0374-1G
1,8-Octanedithiol ≥97.0% (by GC, titration analysis)
1,8-Octanedithiol
CAS Number: 1191-62-4
MDL Number: MFCD00003574
Molecular Formula: C8H18S2
Molecular Weight: 178.35
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 269
Melting point (°C): 1
Flash Point (°C): 110
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,8-Dimercaptooctane
Formula:
C₈H₁₈S₂ MW: 178.36 g/mol Boiling Pt: 269 °C Melting Pt: 1 °C Density: 0.97 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00003574 CAS Number: 1191-62-4 EINECS: 214-738-8 UN: 3334 ADR: 9, |
Specification Test Results
Appearance | Colourless to Almost Colourless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Iodometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...