2-n-Octyl-4-isothiazolin-3-one ≥98.0% (by GC)

Supplier: TCI America

O0343-25G O0343-5G
TCO0343-25GEA 226.05 USD
TCO0343-25G TCO0343-5G
2-n-Octyl-4-isothiazolin-3-one ≥98.0% (by GC)
2-n-Octyl-4-isothiazolin-3-one

CAS Number: 26530-20-1
MDL Number: MFCD00072473
Molecular Formula: C11H19NOS
Molecular Weight: 213.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 142
Freezing point (°C): 23


Product Categories: Bioscience, Fungicide Ingredients
Other Product Names / Synonyms: Octhilinone

Formula: C₁₁H₁₉NOS
MW: 213.34 g/mol
Boiling Pt: 142 °C
MDL Number: MFCD00072473
CAS Number: 26530-20-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=428c840c8099f7c5f98bc8de867ec8daa1d1c853&vid=428c840c8099f7c5f98bc8de867ec8daa1d1c853&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR