1-n-Octyl-2-pyrrolidone ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-Octylpyrrolidin-2-one

O0325-25G O0325-500G O0325-25ML O0325-500ML
TCO0325-25GEA 18.55 USD
TCO0325-25G TCO0325-500G TCO0325-25ML TCO0325-500ML
1-n-Octyl-2-pyrrolidone ≥98.0% (by GC)
1-n-Octyl-2-pyrrolidone

CAS Number: 2687-94-7
Molecular Formula: C12H23NO
Molecular Weight: 197.32
Form: Liquid
Color: Colorless

Formula: C₁₂H₂₃NO
MW: 197.322 g/mol
Boiling Pt: 172 °C
Melting Pt: –23 °C
Flash Pt: 142 °C
MDL Number: MFCD00074965
CAS Number: 2687-94-7
EINECS: 403-700-8
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Melting point (°C) -23
Flash Point (°C) 142
Specific Gravity (20/20) 0.92

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR