Potassium hydrogen monopersulfate >45%

Supplier: TCI America

Synonyms: KMPS, Potassium peroxomonosulphate, Oxone, PMPS, Potassium monopersulphate, Potassium monopersulphate triple salt

O0310-25G O0310-500G
TCO0310-25GEA 35.6 USD
TCO0310-25G TCO0310-500G
Potassium hydrogen monopersulfate >45%
Potassium hydrogen monopersulfate

CAS Number: 37222-66-5
Molecular Formula: H3K5O18S4
Molecular Weight: 614.74
Form: Crystal
Color: White


Other Product Names / Synonyms: Oxone

Formula: 2KHSO₅·KHSO₄·K₂SO₄
MW: 614.76 g/mol
MDL Number: MFCD00040551
CAS Number: 70693-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e942330c822b9e76a39c55b0c4ed117b0c70eb06&vid=f3087f4e921eee45e380584c080f71a67cfccd66&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR