Octanoyl chloride ≥99.0% (by GC)

Supplier: TCI America

O0039-25ML O0039-500ML O0039-100ML
TCO0039-25MLEA 29.32 USD
TCO0039-25ML TCO0039-500ML TCO0039-100ML
Octanoyl chloride ≥99.0% (by GC)
Octanoyl chloride

CAS Number: 111-64-8
MDL Number: MFCD00000766
Molecular Formula: C8H15ClO
Molecular Weight: 162.66
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Melting point (°C): -63
Flash Point (°C): 85
Specific Gravity (20/20): 0.95

Formula: C₈H₁₅ClO
MW: 162.66 g/mol
Boiling Pt: 195 °C
Melting Pt: –63 °C
Density: 0.95 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00000766
CAS Number: 111-64-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba03a51a0a5dd7eb42982f69c1db396de0701063&vid=ba03a51a0a5dd7eb42982f69c1db396de0701063&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR