2-Naphthylacetonitrile ≥98.0% (by GC)
Supplier: TCI America
N0874-25G
N0874-5G
TCN0874-25GEA
202
USD
TCN0874-25G
TCN0874-5G
2-Naphthylacetonitrile ≥98.0% (by GC)
2-Naphthylacetonitrile
CAS Number: 7498-57-9
MDL Number: MFCD00004122
Molecular Formula: C12H9N
Molecular Weight: 167.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 303
Melting point (°C): 86
Other Product Names / Synonyms: 2-Naphthylmethylcyanide
Formula:
C₁₂H₉N MW: 167.21 g/mol Boiling Pt: 303 °C Melting Pt: 86 °C |
MDL Number:
MFCD00004122 CAS Number: 7498-57-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...