Nickel(II) trifluoromethanesulfonate ≥96.0%
Supplier: TCI America
Synonyms:
Trifluoromethanesulphonic acid Nickel(II) Salt, Nickel(II) Triflate, Nickel(II) trifluoromethanesulphonate, Nickel triflate
N0861-1G
N0861-5G
TCN0861-1GEA
37
USD
TCN0861-1G
TCN0861-5G
Nickel(II) trifluoromethanesulfonate ≥96.0%
Nickel(II) trifluoromethanesulfonate
CAS Number: 60871-84-3
MDL Number: MFCD00673740
Molecular Formula: C2F6NiO6S2
Molecular Weight: 356.82
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid Nickel(II) Salt, Nickel(II) Triflate
Formula:
C₂F₆NiO₆S₂ MW: 356.82 g/mol |
MDL Number:
MFCD00673740 CAS Number: 60871-84-3 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...