4-Nitrophenyl isothiocyanate ≥99.0% (by GC)
Supplier: TCI America
N0826-25G
N0826-5G
TCN0826-25GEA
253.2
USD
TCN0826-25G
TCN0826-5G
4-Nitrophenyl isothiocyanate ≥99.0% (by GC)
4-Nitrophenyl isothiocyanate
CAS Number: 2131-61-5
MDL Number: MFCD00007307
Molecular Formula: C7H4N2O2S
Molecular Weight: 180.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 138
Melting point (°C): 112
Other Product Names / Synonyms: Isothiocyanic Acid 4-Nitrophenyl Ester
Formula:
C₇H₄N₂O₂S MW: 180.19 g/mol Boiling Pt: 138 °C Melting Pt: 112 °C |
MDL Number:
MFCD00007307 CAS Number: 2131-61-5 UN: 2923 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...