1,5-Naphthalenedisulfonic acid tetrahydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Naphthalene-1,5-disulphonic acid tetrahydrate

N0763-25G N0763-500G
TCN0763-25GEA 36.88 USD
TCN0763-25G TCN0763-500G
1,5-Naphthalenedisulfonic acid tetrahydrate ≥98.0% (by HPLC, titration analysis)
1,5-Naphthalenedisulfonic acid tetrahydrate

CAS Number: 211366-30-2
Molecular Formula: C10H8O6S2
Molecular Weight: 288.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₁₆O₁₀S₂
MW: 360.36 g/mol
MDL Number: MFCD00149265
CAS Number: 211366-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=017591df69abc4ce5bd855089e88b762081947d9&vid=017591df69abc4ce5bd855089e88b762081947d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR