4-Nitrophenylsulfur pentafluoride ≥96.0% (by GC)
Supplier: TCI America
N0743-1G
N0743-5G
TCN0743-1GEA
26
USD
TCN0743-1G
TCN0743-5G
4-Nitrophenylsulfur pentafluoride ≥96.0% (by GC)
4-Nitrophenylsulphur pentafluoride
CAS Number: 2613-27-6
MDL Number: MFCD00221619
Molecular Formula: C6H4F5NO2S
Molecular Weight: 249.16
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 77
Melting point (°C): 39
Formula:
C₆H₄F₅NO₂S Boiling Pt: 77 °C Melting Pt: 39 °C |
MDL Number:
MFCD00221619 CAS Number: 2613-27-6 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...