Perfluorobutane-1-sulfonic acid ≥98.0%
Supplier: TCI America
Synonyms:
Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid
N0709-25G
N0709-5G
TCN0709-25GEA
227.87
USD
TCN0709-25G
TCN0709-5G
Perfluorobutane-1-sulfonic acid ≥98.0%
Perfluorobutane-1-sulphonic acid
CAS Number: 375-73-5
MDL Number: MFCD01320794
Molecular Formula: C4HF9O3S
Molecular Weight: 300.09
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 212
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Perfluoro-1-butanesulfonic Acid
Formula:
C₄HF₉O₃S Boiling Pt: 212 °C |
MDL Number:
MFCD01320794 CAS Number: 375-73-5 UN: 3265 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...