4-Methyl-3-nitrobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Methyl-3-nitrobenzene-1-sulphonyl chloride
N0704-25G
N0704-5G
TCN0704-25GEA
1002
USD
TCN0704-25G
TCN0704-5G
4-Methyl-3-nitrobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
4-Methyl-3-nitrobenzenesulphonyl chloride
CAS Number: 616-83-1
MDL Number: MFCD00129811
Molecular Formula: C7H6ClNO4S
Molecular Weight: 235.64
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 36
Other Product Names / Synonyms: 3-Nitro-p-toluenesulfonyl Chloride
Formula:
C₇H₆ClNO₄S MW: 235.65 g/mol Boiling Pt: 154 °C Melting Pt: 36 °C |
MDL Number:
MFCD00129811 CAS Number: 616-83-1 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...