3-Nitro-o-cresol ≥98.0%

Supplier: TCI America

Synonyms: 2-Methyl-3-nitrophenol, 1-Hydroxy-2-methyl-3-nitrobenzene

N0627-25G N0627-5G
TCN0627-25GEA 71.1 USD
TCN0627-25G TCN0627-5G
3-Nitro-o-cresol ≥98.0%
3-Nitro-o-cresol

CAS Number: 5460-31-1
MDL Number: MFCD00007241
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152


Other Product Names / Synonyms: 2-Hydroxy-6-nitrotoluene, 2-Methyl-3-nitrophenol

Formula: C₇H₇NO₃
Melting Pt: 152 °C
MDL Number: MFCD00007241
CAS Number: 5460-31-1
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffd4881cc438070ca80cb63544811fa0e79c1069&vid=ffd4881cc438070ca80cb63544811fa0e79c1069&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR