α-Bromo-3-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α-Bromo-m-nitrotoluene, 1-(Bromomethyl)-3-nitrobenzene, 3-Nitrobenzyl bromide, m-Nitrobenzyl bromide

N0625-10G N0625-25G
TCN0625-10GEA 16.28 USD
TCN0625-10G TCN0625-25G
α-Bromo-3-nitrotoluene ≥98.0% (by GC)
α-Bromo-3-nitrotoluene

CAS Number: 3958-57-4
MDL Number: MFCD00007271
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 58


Other Product Names / Synonyms: alpha-Bromo-3-nitrotoluene

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 115 °C
Melting Pt: 58 °C
MDL Number: MFCD00007271
CAS Number: 3958-57-4
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c70587e5f0602af2b1226a6870432fdca12993a1&vid=c78b14295b13d147b63adf8910d7ead7cfcf8790&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR