Norcamphor ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Oxonorbornane, Bicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]-2-heptanone, 2,5-Methanocyclohexanone, 2-Norbornanone

N0511-25G N0511-5G
TCN0511-25GEA 376 USD
TCN0511-25G TCN0511-5G
Norcamphor ≥98.0% (by GC)
Norcamphor

CAS Number: 497-38-1
MDL Number: MFCD00074823
Molecular Formula: C7H10O
Molecular Weight: 110.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 55
Melting point (°C): 95
Flash Point (°C): 33


Other Product Names / Synonyms: Bicyclo[2.2.1]-2-heptanone, 2-Norcamphor

Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 55 °C
Melting Pt: 95 °C
Flash Pt: 33 °C
MDL Number: MFCD00074823
CAS Number: 497-38-1
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b52e990ef37a1b1d25b3b948054ec5089738e954&vid=b52e990ef37a1b1d25b3b948054ec5089738e954&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR