ω-Bromo-4-nitroacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone

N0473-25G N0473-5G
TCN0473-25GEA 33.5 USD
TCN0473-25G TCN0473-5G
ω-Bromo-4-nitroacetophenone ≥98.0% (by GC, titration analysis)
ω-Bromo-4-nitroacetophenone

CAS Number: 99-81-0
MDL Number: MFCD00007356
Molecular Formula: C8H6BrNO3
Molecular Weight: 244.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 98


Other Product Names / Synonyms: 4-Nitrophenacyl Bromide

Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00007356
CAS Number: 99-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c88f5b53c4ca0399c4faeb9186f22334d645a47&vid=2c88f5b53c4ca0399c4faeb9186f22334d645a47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR