3-Nitro-o-toluidine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Nitro-o-methylaniline, 3-Nitro-2-methylaniline, 2-Methyl-3-nitroaniline, 2-Amino-6-nitrotoluene

N0455-250G N0455-25G
TCN0455-250GEA 772 USD
TCN0455-250G TCN0455-25G
3-Nitro-o-toluidine ≥98.0% (by GC)
3-Nitro-o-toluidine

CAS Number: 603-83-8
MDL Number: MFCD00007731
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 305
Melting point (°C): 90


Other Product Names / Synonyms: 1-Amino-2-methyl-3-nitrobenzene, 3-Nitro-o-toluidine

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 305 °C
Melting Pt: 90 °C
MDL Number: MFCD00007731
CAS Number: 603-83-8
UN: 2660
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f0283c15e3712ca69ea009a3cb6260593e09a40&vid=1f0283c15e3712ca69ea009a3cb6260593e09a40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR