α-Bromo-2-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide

N0404-25G N0404-5G
TCN0404-25GEA 140 USD
TCN0404-25G TCN0404-5G
α-Bromo-2-nitrotoluene ≥98.0% (by GC)
α-Bromo-2-nitrotoluene

CAS Number: 3958-60-9
MDL Number: MFCD00007184
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis
Other Product Names / Synonyms: alpha-Bromo-2-nitrotoluene

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00007184
CAS Number: 3958-60-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=356c7441750d555a5df2ea5d652cdad641b87be1&vid=41bba3427b4c0cb48c2be8f1a3fa8e7b9974d958&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR