3-Nitrophenylhydrazinium chloride

Supplier: TCI America

Synonyms: 3-Nitrophenylhydrazine hydrochloride, 1-(3-Nitrophenyl)hydrazine hydrochloride, (3-Nitrophenyl)hydrazine hydrochloride

N0232-25G N0232-5G
TCN0232-25GEA 140 USD
TCN0232-25G TCN0232-5G
3-Nitrophenylhydrazinium chloride
3-Nitrophenylhydrazinium chloride

CAS Number: 636-95-3
MDL Number: MFCD00012939
Molecular Formula: C6H7N3O2
Molecular Weight: 189.60
Form: Crystal
Color: Pale Yellow

Formula: C₆H₈ClN₃O₂
MDL Number: MFCD00012939
CAS Number: 636-95-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55c2ee8510870e3534cb6957af95796dd3870735&vid=55c2ee8510870e3534cb6957af95796dd3870735&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR