3-Nitro-p-cresol ≥98.0%

Supplier: TCI America

Synonyms: 4-Methyl-3-nitrophenol, 1-Hydroxy-4-methyl-3-nitrobenzene

N0210-25G N0210-5G
TCN0210-25GEA 54 USD
TCN0210-25G TCN0210-5G
3-Nitro-p-cresol ≥98.0%
3-Nitro-p-cresol

CAS Number: 2042-14-0
MDL Number: MFCD00007244
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 77


Other Product Names / Synonyms: 4-Hydroxy-1-methyl-2-nitrobenzene, 4-Methyl-3-nitrophenol

Formula: C₇H₇NO₃
Melting Pt: 77 °C
MDL Number: MFCD00007244
CAS Number: 2042-14-0
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f765c4eeb2b8611f40f2cfd5ee4d816484fefed&vid=5f765c4eeb2b8611f40f2cfd5ee4d816484fefed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR