2-Nitrobenzyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-(Chloromethyl)-2-nitrobenzene, 2-Nitrobenzyl chloride, α-Chloro-o-nitrotoluene, alpha-Chloro-2-nitrotoluene
N0182-25G
N0182-5G
TCN0182-25GEA
210
USD
TCN0182-25G
TCN0182-5G
2-Nitrobenzyl chloride ≥98.0% (by GC, titration analysis)
2-Nitrobenzyl chloride
CAS Number: 612-23-7
MDL Number: MFCD00007185
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 49
Flash Point (°C): 112
Other Product Names / Synonyms: alpha-Chloro-2-nitrotoluene
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Boiling Pt: 133 °C Melting Pt: 49 °C Flash Pt: 112 °C |
MDL Number:
MFCD00007185 CAS Number: 612-23-7 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...