4-Nitrobenzonitrile ≥98.0%

Supplier: TCI America

N0172-250G N0172-25G
TCN0172-250GEA 240 USD
TCN0172-250G TCN0172-25G
4-Nitrobenzonitrile ≥98.0%
4-Nitrobenzonitrile

CAS Number: 619-72-7
MDL Number: MFCD00007279
Molecular Formula: C7H4N2O2
Molecular Weight: 148.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147

Formula: C₇H₄N₂O₂
Melting Pt: 147 °C
MDL Number: MFCD00007279
CAS Number: 619-72-7
UN: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95cdae93ed7ef073b5f8458372764a2001cc439f&vid=df38d507067f0c104be91a49851deb286c0931ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR