4-Nitrobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Nitrobenzene-1-sulphonyl chloride
N0144-25G
N0144-5G
TCN0144-25GEA
45.85
USD
TCN0144-25G
TCN0144-5G
4-Nitrobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
4-Nitrobenzenesulphonyl chloride
CAS Number: 98-74-8
MDL Number: MFCD00007445
Molecular Formula: C6H4ClNO4S
Molecular Weight: 221.61
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 181
Melting point (°C): 79
Formula:
C₆H₄ClNO₄S MW: 221.62 g/mol Boiling Pt: 181 °C Melting Pt: 79 °C |
MDL Number:
MFCD00007445 CAS Number: 98-74-8 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...