Methyl-2-(chloromethyl)benzoate ≥97.0% (by GC)

Supplier: TCI America

M2203-25G M2203-5G
TCM2203-25GEA 640 USD
TCM2203-25G TCM2203-5G
Methyl-2-(chloromethyl)benzoate ≥97.0% (by GC)
Methyl 2-(chloromethyl)benzoate

CAS Number: 34040-62-5
MDL Number: MFCD06200781
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: 2-(Chloromethyl)benzoic Acid Methyl Ester

Formula: C₉H₉ClO₂
Density: 1.23 (20 °C)
MDL Number: MFCD06200781
CAS Number: 34040-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=363f5303732f3703f91b5fc7286e40582148e93b&vid=363f5303732f3703f91b5fc7286e40582148e93b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR