Methyl-2-bromo-2-(2-chlorophenyl)acetate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: alpha-Bromo-2-chlorophenylacetic acid methyll ester, Methyl α-Bromo-2-chlorophenylacetate, Methyl alpha-Bromo-2-chlorophenylacetate

M2175-25G M2175-5G
TCM2175-25GEA 205 USD
TCM2175-25G TCM2175-5G
Methyl-2-bromo-2-(2-chlorophenyl)acetate ≥98.0% (by GC, titration analysis)
Methyl-2-bromo-2-(2-chlorophenyl)acetate

CAS Number: 85259-19-4
MDL Number: MFCD08444353
Molecular Formula: C9H8BrClO2
Molecular Weight: 263.52
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.56
Storage Temperature: 0-10°C


Other Product Names / Synonyms: alpha-Bromo-2-chlorophenylacetic Acid Methyl Ester

Formula: C₉H₈BrClO₂
MW: 263.5156 g/mol
Density: 1.56 (20 °C)
MDL Number: MFCD08444353
CAS Number: 85259-19-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e8b347f99f93fafb788db508099f665380ed41&vid=d4e8b347f99f93fafb788db508099f665380ed41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR