(R)-(-)-1-Methoxy-2-propanol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(R)-1-Methoxy-2-propanol, (R)-(-)-Propylene glycol 1-Monomethyl ether
M2166-25G
M2166-5G
TCM2166-25GEA
124
USD
TCM2166-25G
TCM2166-5G
(R)-(-)-1-Methoxy-2-propanol ≥98.0% (by GC)
(R)-(-)-1-Methoxy-2-propanol
CAS Number: 4984-22-9
MDL Number: MFCD01632587
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 32
Specific Gravity (20/20): 0.92
Specific rotation [a]20/D: -22 deg (C=10, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-Propylene Glycol 1-Monomethyl Ether
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 120 °C Density: 0.92 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD01632587 CAS Number: 4984-22-9 UN: 3092 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...