O-(2-Methoxyisopropyl)hydroxylamine ≥95.0% (by GC, titration analysis)

Supplier: TCI America

M2085-1G M2085-5G
TCM2085-1GEA 148 USD
TCM2085-1G TCM2085-5G
O-(2-Methoxyisopropyl)hydroxylamine ≥95.0% (by GC, titration analysis)
O-(2-Methoxyisopropyl)hydroxylamine

CAS Number: 103491-33-4 Molecular Formula: C4H11NO2 Molecular Weight: 105.14 Purity/Analysis Method: 95.0% (GC,T) Form: Clear Liquid Boiling point (°C): 73 Specific Gravity (20/20): 0.98 Storage Temperature: 0-10°C

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 73 °C
Density: 0.98 (20 °C)
CAS Number: 103491-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b07c434e05b375c9600f7cf9b223a7c1e0b6390f&vid=b07c434e05b375c9600f7cf9b223a7c1e0b6390f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR