3-Methoxy-4-methylbenzonitrile ≥98.0% (by GC)

Supplier: TCI America

M1977-1G M1977-5G
TCM1977-1GEA 25 USD
TCM1977-1G TCM1977-5G
3-Methoxy-4-methylbenzonitrile ≥98.0% (by GC)
2-Methoxy-5-methylbenzonitrile

CAS Number: 3556-60-3
MDL Number: MFCD03428590
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54


Other Product Names / Synonyms: 4-Cyano-2-methoxytoluene, 5-Cyano-2-methylanisole

Formula: C₉H₉NO
Melting Pt: 54 °C
MDL Number: MFCD03428590
CAS Number: 3556-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d496ea81ea1ef3a7faff6b1b4bab6f0714dd98af&vid=d496ea81ea1ef3a7faff6b1b4bab6f0714dd98af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR