3,4-(Methylenedioxy)phenylacetonitrile ≥98.0% (by GC)

Supplier: TCI America

M1923-25G M1923-5G
TCM1923-25GEA 291 USD
TCM1923-25G TCM1923-5G
3,4-(Methylenedioxy)phenylacetonitrile ≥98.0% (by GC)
3,4-(Methylenedioxy)phenylacetonitrile

CAS Number: 4439-02-5
MDL Number: MFCD00005835
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 43
Flash Point (°C): 110


Other Product Names / Synonyms: 3,4-Methylenedioxybenzyl Cyanide, Homopiperonylonitrile, 1,3-Benzodioxole-5-acetonitrile

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 140 °C
Melting Pt: 43 °C
Flash Pt: 110 °C
MDL Number: MFCD00005835
CAS Number: 4439-02-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35fe140cee1ece310aa5592c0c13a07809c404e4&vid=35fe140cee1ece310aa5592c0c13a07809c404e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR