Methyl-3-chloro-2,2,3,3-tetrafluoropropionate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 3-Chloro-2,2,3,3-tetrafluoropropionic Acid Methyl Ester

M1902-1G M1902-5G
TCM1902-1GEA 118 USD
TCM1902-1G TCM1902-5G
Methyl-3-chloro-2,2,3,3-tetrafluoropropionate ≥97.0% (by GC)
Methyl-3-chloro-2,2,3,3-tetrafluoropropionate

CAS Number: 127589-63-3
MDL Number: MFCD00155715
Molecular Formula: C4H3ClF4O2
Molecular Weight: 194.51
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.46


Other Product Names / Synonyms: 3-Chloro-2,2,3,3-tetrafluoropropionic Acid Methyl Ester

Formula: C₄H₃ClF₄O₂
MW: 194.51 g/mol
Density: 1.46 (20 °C)
MDL Number: MFCD00155715
CAS Number: 127589-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fbf1ff698a34c04b53aff48d828e270c369f9d8&vid=7fbf1ff698a34c04b53aff48d828e270c369f9d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR