3-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(3-Methoxyphenyl)ethylamine
M1891-25G
M1891-5G
TCM1891-25GEA
167.4
USD
TCM1891-25G
TCM1891-5G
3-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)
3-Methoxyphenethylamine
CAS Number: 2039-67-0
MDL Number: MFCD00008187
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 110
Specific Gravity (20/20): 1.04
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: 3-(2-Aminoethyl)anisole, 3-Methoxyphenethylamine
Formula:
C₉H₁₃NO MW: 151.21 g/mol Boiling Pt: 119 °C Density: 1.04 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008187 CAS Number: 2039-67-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...