3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate ≥95.0%
Supplier: TCI America
Synonyms:
3-Methoxy-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulphonic acid 3-Methoxy-2-(trimethylsilyl)phenyl Ester
CAS Number: 217813-03-1
Molecular Formula: C11H15F3O4SSi
Molecular Weight: 328.38
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.25
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Benzyne Precursors (Multi-component Reaction), Silicon Compounds (for Synthesis), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 3-Methoxy-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 3-Methoxy-2-(trimethylsilyl)phenyl Ester
Formula:
C₁₁H₁₅F₃O₄SSi Density: 1.25 (20 °C) |
MDL Number:
MFCD10566916 CAS Number: 217813-03-1 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...