2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 2-Methyl-6-(trimethylsilyl)phenyl Triflate, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulphonate

M1883-1G M1883-5G
TCM1883-1GEA 205 USD
TCM1883-1G TCM1883-5G
2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate ≥96.0% (by GC)
2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulfonate

CAS Number: 556812-44-3
Molecular Formula: C11H15F3O3SSi
Molecular Weight: 312.38
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Benzyne Precursors (Multi-component Reaction), Silicon Compounds (for Synthesis), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Methyl-6-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-Methyl-6-(trimethylsilyl)phenyl Ester, 6-(Trimethylsilyl)-o-tolyl Trifluoromethanesulfonate

Formula: C₁₁H₁₅F₃O₃SSi
MW: 312.38 g/mol
Density: 1.23 (20 °C)
CAS Number: 556812-44-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb21d1e97911d423177e7f22c5f93ed8cca8dd4e&vid=eb21d1e97911d423177e7f22c5f93ed8cca8dd4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR