4-Methyl-2-(trimethylsilyl)phenyl trifluoromethanesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
2-(Trimethylsilyl)-p-tolyl Trifluoromethanesulphonate, Trifluoromethanesulphonic acid 4-Methyl-2-(trimethylsilyl)phenyl Ester, 4-Methyl-2-(trimethylsilyl)phenyl Trifluoromethanesulphonate, 4-Methyl-2-(trimethylsilyl)phenyl Triflate
CAS Number: 262373-15-9
Molecular Formula: C11H15F3O3SSi
Molecular Weight: 312.38
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.22
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Benzyne Precursors (Multi-component Reaction), Silicon Compounds (for Synthesis), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 4-Methyl-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 4-Methyl-2-(trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)-p-tolyl Trifluoromethanesulfonate
Formula:
C₁₁H₁₅F₃O₃SSi MW: 312.38 g/mol Density: 1.22 (20 °C) |
MDL Number:
MFCD10566914 CAS Number: 262373-15-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...