2-Methyl-3-(propyldithio)furan ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Methyl-3-(propyldisulphanyl)furan, 2-Methyl-3-furyl Propyl Disulphide

M1840-1G M1840-5G
TCM1840-1GEA 34.5 USD
TCM1840-1G TCM1840-5G
2-Methyl-3-(propyldithio)furan ≥97.0% (by GC)
2-Methyl-3-(propyldithio)furan

CAS Number: 61197-09-9
MDL Number: MFCD01729816
Molecular Formula: C8H12OS2
Molecular Weight: 188.30
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Methyl-3-furyl Propyl Disulfide

Formula: C₈H₁₂OS₂
MW: 188.30 g/mol
Density: 1.10 (20 °C)
MDL Number: MFCD01729816
CAS Number: 61197-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e25bebf575f3225cf590477ee6d3e46144a7936&vid=1e25bebf575f3225cf590477ee6d3e46144a7936&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR