2-Methyl-3-(methylthio)furan ≥98.0% (by GC)
Supplier: TCI America
M1838-25G
M1838-5G
TCM1838-25GEA
222
USD
TCM1838-25G
TCM1838-5G
2-Methyl-3-(methylthio)furan ≥98.0% (by GC)
2-Methyl-3-(methylthio)furan
CAS Number: 63012-97-5
MDL Number: MFCD03701635
Molecular Formula: C6H8OS
Molecular Weight: 128.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 59
Specific Gravity (20/20): 1.07
Formula:
C₆H₈OS MW: 128.19 g/mol Boiling Pt: 132 °C Density: 1.07 (20 °C) Flash Pt: 59 °C |
MDL Number:
MFCD03701635 CAS Number: 63012-97-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...