2-Methyl-3-(methyldithio)furan ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Methyl-3-(methyldisulphanyl)furan, Methyl 2-Methyl-3-furyl Disulphide
M1837-1G
M1837-5G
TCM1837-1GEA
32
USD
TCM1837-1G
TCM1837-5G
2-Methyl-3-(methyldithio)furan ≥98.0% (by GC)
2-Methyl-3-(methyldithio)furan
CAS Number: 65505-17-1
MDL Number: MFCD00038643
Molecular Formula: C6H8OS2
Molecular Weight: 160.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 37
Flash Point (°C): 84
Specific Gravity (20/20): 1.17
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Methyl 2-Methyl-3-furyl Disulfide
Formula:
C₆H₈OS₂ MW: 160.25 g/mol Boiling Pt: 37 °C Density: 1.17 (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00038643 CAS Number: 65505-17-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...