10-Methyl-9-phenylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 10-Methyl-9-phenylacridin-10-ium perchlorate

M1775-1G M1775-5G
TCM1775-1GEA 114 USD
TCM1775-1G TCM1775-5G
10-Methyl-9-phenylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
10-Methyl-9-phenylacridinium perchlorate

CAS Number: 36519-61-6
Molecular Formula: C20H16ClNO4
Molecular Weight: 369.80
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₂₀H₁₆ClNO₄
MW: 369.80 g/mol
MDL Number: MFCD09038537
CAS Number: 36519-61-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d52640dbb82f5840756d089cdb3ac114a59bb5bc&vid=d52640dbb82f5840756d089cdb3ac114a59bb5bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR