9-Mesityl-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 10-Methyl-9-(2,4,6-trimethylphenyl)acridinium Perchlorate

M1774-1G M1774-5G
TCM1774-1GEA 154.62 USD
TCM1774-1G TCM1774-5G
9-Mesityl-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
9-Mesityl-10-methylacridinium perchlorate

CAS Number: 674783-97-2
Molecular Formula: C23H22ClNO4
Molecular Weight: 411.88
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Oxidation, Oxidation, Oxidation
Other Product Names / Synonyms: 10-Methyl-9-(2,4,6-trimethylphenyl)acridinium Perchlorate

Formula: C₂₃H₂₂ClNO₄
MW: 411.88 g/mol
CAS Number: 674783-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cccda50c3c782dea1168bc4f34dd60a8d099ed8&vid=0cccda50c3c782dea1168bc4f34dd60a8d099ed8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR