(R)-(+)-β-Methylphenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-(+)-beta-Methylphenethylamine, (R)-(+)-2-Phenyl-1-propylamine

M1746-25G M1746-5G
TCM1746-25GEA 234.03 USD
TCM1746-25G TCM1746-5G
(R)-(+)-β-Methylphenethylamine ≥98.0% (by GC, titration analysis)
(R)-(+)-β-Methylphenethylamine

CAS Number: 28163-64-6
MDL Number: MFCD00216740
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 81
Specific Gravity (20/20): 0.95
Specific rotation [a]20/D: 32 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-2-Phenyl-1-propylamine

Formula: C₉H₁₃N
Boiling Pt: 102 °C
Density: 0.95 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00216740
CAS Number: 28163-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ceacb6d4e186abaa584dc4a3ca8fd1271643c0&vid=58ceacb6d4e186abaa584dc4a3ca8fd1271643c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR